4-Amino-7-chloroquinoline

Synonyms: 7-Chloroquinolin-4-amine

CFF545.0.5 CFF545.1
CHMPCFF545.0.5EA 0 GBP
CHMPCFF545.0.5 CHMPCFF545.1
4-Amino-7-chloroquinoline
4-Amino-7-chloroquinoline
Formula: C₉H₇ClN₂
MW: 178,6183 g/mol
MDL Number: MFCD00828822
CAS Number: 1198-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR