6-Bromo-4-chloroquinoline 96%

Supplier: Thermo Scientific
Warning

H54902.MD H54902.03
H54902.MDEA 108 GBP
H54902.MD H54902.03
6-Bromo-4-chloroquinoline 96%
6-Bromo-4-chloroquinoline

Purity: 96% CAS No.: 65340-70-7 Molecular Formula: C9H5BrClN Formula Weight: 242.50 Storage Temperature: 2°C to 8°C Melting Point: 110-112° MDL No.: MFCD00511001

Formula: C₉H₅BrClN
MW: 242,50 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD00511001
CAS Number: 65340-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR