3-Benzyloxy-2-fluorophenylboronic acid pinacol ester 96%
Supplier: Thermo Scientific
Synonyms:
2-(3-(Benzyloxy)-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Benzyloxy-2-fluorobenzeneboronic acid pinacol ester
H59049.MD
H59049.03
H59049.MDEA
124
GBP
H59049.MD
H59049.03
3-Benzyloxy-2-fluorophenylboronic acid pinacol ester 96%
3-Benzyloxy-2-fluorophenylboronic acid pinacol ester
Formula:
C₁₉H₂₂BFO₃ MW: 328,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22419264 CAS Number: 874152-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...