2,5-Bis(trifluoromethyl)thiobenzamide ≥97%

Supplier: Thermo Scientific
Warning

H33419.MD H33419.03
H33419.MDEA 60.1 GBP
H33419.MD H33419.03
2,5-Bis(trifluoromethyl)thiobenzamide ≥97%
2,5-Bis(trifluoromethyl)thiobenzamide

Purity: 97% CAS No.: 1186194-57-9 Molecular Formula: C9H5F6NS Formula Weight: 273.20 Melting Point: 84-88° MDL No.: MFCD09757524

Formula: C₉H₅F₆NS
MW: 273,20 g/mol
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD09757524
CAS Number: 1186194-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR