Methyl-2-amino-3-bromo-5-methoxybenzoate 96%

Supplier: Thermo Scientific
Warning

H54844.MD H54844.03
H54844.MDEA 117 GBP
H54844.MD H54844.03
Methyl-2-amino-3-bromo-5-methoxybenzoate 96%
Methyl 2-amino-3-bromo-5-methoxybenzoate

Purity: 96% Molecular Formula: C9H10BrNO3 Formula Weight: 260.09 MDL No.: MFCD20527215

Formula: C₉H₁₀BrNO₃
MW: 260,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20527215
CAS Number: 1378874-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR