4-(Diethylcarbamoyl)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-(Diethylcarbamoyl)benzeneboronic acid, 4-(N,N-Diethylaminocarbonyl)phenylboronic acid

H52986.03 H52986.06
H52986.03EA 26.1 GBP
H52986.03 H52986.06
4-(Diethylcarbamoyl)phenylboronic acid ≥98%
4-(Diethylcarbamoyl)phenylboronic acid

Purity: 98% CAS No.: 389621-80-1 Molecular Formula: C11H16BNO3 Formula Weight: 221.06 Melting Point: 138-142° MDL No.: MFCD03411949

Formula: C₁₁H₁₆BNO₃
MW: 221,06 g/mol
Melting Pt: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD03411949
CAS Number: 389621-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR