Triallylphosphine ≥97%

Supplier: Thermo Scientific

H27881.06 H27881.03
H27881.06EA 0 GBP
H27881.06 H27881.03
Triallylphosphine ≥97%
Triallylphosphine
Formula: C₉H₁₅P
MW: 154,189 g/mol
MDL Number: MFCD00014956
CAS Number: 16523-89-0
EINECS: 240-591-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR