Dimethylzinc 1.2 mol/l toluene, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

377241000 377248000
377241000.EA 531 GBP
377241000. 377248000.
Dimethylzinc 1.2 mol/l toluene, AcroSeal®
Dimethylzinc
Formula: C₂H₆Zn
MW: 95.478 g/mol
Density: 0.93 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00014854
CAS Number: 544-97-8
EINECS: 208-884-1
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Concentration 11.5 to 12.5 wt% (On Zn basis)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR