3-Chloroimidazo[1,2-a]pyridine-6-carboxylic acid 95%

Supplier: Thermo Fisher Scientific
Warning

H35089.MD H35089.03
H35089.MDEA 0 GBP
H35089.MD H35089.03
3-Chloroimidazo[1,2-a]pyridine-6-carboxylic acid 95%
3-Chloroimidazo[1,2-a]pyridine-6-carboxylic acid
Formula: C₈H₅ClN₂O₂
Melting Pt: ∼ 300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD07021510
CAS Number: 900019-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR