Ethanol eurodenatured ≥91.5%, GPR RECTAPUR® (denatured with 3% MEK and 3% IPA and 12 ppm denatonium benzoate)
Supplier: VWR Chemicals
|
Danger
|
84835.360EA
0
GBP
84835.360
84835.290
84835.410
84835.320
Ethanol eurodenatured ≥91.5%, GPR RECTAPUR® (denatured with 3% MEK and 3% IPA and 12 ppm denatonium benzoate)
Ethanol Eurodenatured
Completely denatured alcohol (CDA).
Denatonium benzoate (a tasting agent) is also known by several trade names including Bitrex, BITTERANT-b, BITTER+PLUS.
Formula:
H₃CCH₂OH Boiling Pt: 78 °C (1013 hPa) Melting Pt: –114 °C Density: 0,79 g/cm³ (20 °C) Flash Pt: 16,6 °C (closed cup) Storage Temperature: Ambient |
CAS Number:
64-17-5 EINECS: 200-578-6 UN: 1170 ADR: 3,II |
Specification Test Results
Assay (V/V) | Min. 91.5 % |
Denatonium benzoate (11,4 -13,4 ppm) | Passes test |
Methyl ethyl ketone (V/V) | 2.55 - 3.11 % |
2-Propanol (V/V) | 2.55 - 3.11 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...