Industrial gloves, Titan 385 / 388 / 391
Supplier: MAPA
34385000 34385009 34391009 34391000 34388009 34388008 34388000 34391008
MAPA34385000EA 55.2 GBP
MAPA34385000 MAPA34385009 MAPA34391009 MAPA34391000 MAPA34388009 MAPA34388008 MAPA34388000 MAPA34391008
Industrial gloves, Titan 385 / 388 / 391
Gloves
These textile gloves either feature a ¾ or a full nitrile coating.
  • Freedom of movement: exceptionally comfortable textile support (no seams on the working surfaces)
  • Excellent resistance to oils and greases
  • Longer service life: quality of textile support and nitrile coating
  • Vented back models are available with or without knitted cuffs
Ideal for waste collection, forestry, metal shearing or cutting and the handling of thick, untrimmed sheet metal (not sharp-edged).
Certifications: EN 388:2016 (4111X); Cat. II