Cut resistant gloves, KryTech 395
Supplier: MAPA
34395320 34395328 34395329
MAPA34395320EA 54.6 GBP
MAPA34395320 MAPA34395328 MAPA34395329
Cut resistant gloves, KryTech 395
Gloves
These multi-purpose nitrile gloves with textile lining combine protection against cuts, liquids and contact heat up to 250 °C.
  • Combined long lasting chemical protection and cut protection
  • Entirely liquid-proof, withstanding mechanical aggression
  • Proven long-lasting performance, measured after 5 washes
  • Good grip for picking up wet and slippery objects safely
Ideal for sheet metal handling, metal rolling, stamping and assembly, cutting aluminium strips, as well as collection and sorting of industrial and household waste.
Certifications: EN 388:2016 (4X43D), EN ISO 374-1 Type B (JKOPT), EN 374-5, EN 407 (X1XXXX), ISO 13997: 20,4N (ANSI Level 3, 2080 gg); Cat. III