Bromocresol purple solution Reag. Ph. Eur. 1012701
Supplier: VWR Chemicals
Synonyms:
3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
, 5',5''-Dibromo-o-cresolsulphonphthalein
87793.180EA
58.4
GBP
87793.180
87793.180_T
VWRC87793.180_DP
Bromocresol purple solution Reag. Ph. Eur. 1012701
Bromocresol purple
Formula:
C₂₁H₁₆Br₂O₅S MW: 540,23 g/mol |
MDL Number:
MFCD00011681 CAS Number: 115-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...