Chlorotrimethylsilane GC, Supelco®
Supplier: Merck
|
Danger
|
Synonyms:
TMCS, TMSC, Trimethylchlorosilane
MQ100
1.02333.0250
1.02333.0250EA
114
GBP
1.02333.0250
Chlorotrimethylsilane GC, Supelco®
Chlorotrimethylsilane
Formula:
C₃H₉ClSi MW: 108.64 g/mol Boiling Pt: 59 Melting Pt: –58 °C Density: 0.8576 g/cm³ (20 °C) Flash Pt: –18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000502 CAS Number: 75-77-4 EINECS: 200-900-5 UN: 1298 ADR: 3 (8) II REACH: 01-2119457596-25 |
Specification Test Results
Solubility | (20 °C) (rigorous decomposition) |
Melting Point | -58 °C |
Molar Mass | 108.64 g/mol |
Boiling Point | 57 °C (1013 hPa) |
Vapor Pressure | 250 hPa (20 °C) |
Flash Point | -18 °C |
Explosion Limit | 1.5 - 46 %(V) |
Density | 0.86 g/cm3 (20 °C) |
Ignition Point | 420 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...