Histamine (free base)
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-(1H-Imidazol-4-yl)ethan-1-amine
J61727.03EA
78.5
GBP
J61727.03
Histamine (free base)
Histamine (free base)
Formula:
C₅H₉N₃ MW: 111,15 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00005210 CAS Number: 51-45-6 EINECS: 200-100-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...