Coconut oil, pure, refined
Supplier: Thermo Scientific
365471000
365475000
ACRO365471000EA
45
GBP
ACRO365471000
ACRO365475000
Coconut oil, pure, refined
Coconut oil
Melting Pt:
23…26 °C Flash Pt: >200 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00130817 CAS Number: 8001-31-8 EINECS: 232-282-8 |
Specification Test Results
Appearance | Colorless to white to light yellow Oily liquid or solid |
Acid value | ≤0.5 mg KOH/g |
Iodine value | 7 to 12 g I2/100 g |
Peroxide value | ≤5 meq O2/kg |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...