4-Phenyl-1-buten-4-ol 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-Phenyl-3-buten-1-ol
380860010
380860050
ACRO380860010EA
53.3
GBP
ACRO380860010
ACRO380860050
4-Phenyl-1-buten-4-ol 97%
4-Phenyl-1-buten-4-ol
Formula:
C₁₀H₁₂O MW: 148,20 g/mol Flash Pt: 103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039617 CAS Number: 936-58-3 |
Specification Test Results
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5290 to 1.5320 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...