2-Amino-5,6-dichlorobenzimidazole 96%

Supplier: Thermo Scientific
Danger

Synonyms: 5,6-Dichloro-1H-benzo[d]imidazol-2-amine

H54597.MD H54597.03
H54597.MDEA 122 GBP
H54597.MD H54597.03
2-Amino-5,6-dichlorobenzimidazole 96%
2-Amino-5,6-dichlorobenzimidazole

Purity: 96% CAS No.: 18672-03-2 Molecular Formula: C7H5Cl2N3 Formula Weight: 202.04 Melting Point: 244-246° MDL No.: MFCD08234602

Formula: C₇H₅Cl₂N₃
MW: 202,04 g/mol
Melting Pt: 244…246 °C
Storage Temperature: Ambient
MDL Number: MFCD08234602
CAS Number: 18672-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR