4-Methylmorpholine 99.5%, AcroSeal®, purified by redistillation

Supplier: Thermo Scientific
Danger

Synonyms: NMM, N-Methylmorpholine

432800010 432801000
432800010.EA 137 GBP
432800010. 432801000.
4-Methylmorpholine 99.5%, AcroSeal®, purified by redistillation
4-Methylmorpholine
Formula: C₅H₁₁NO
MW: 101,15 g/mol
Boiling Pt: 115…116 °C (750 mmHg)
Melting Pt: –65 °C
Density: 0,92 g/cm³
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00006175
CAS Number: 109-02-4
EINECS: 203-640-0
UN: 2535
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4339 to 1.4359 (20°C, 589 nm)
Water ≤0.5 %
GC ≥99.4 %
Color scale ≤20 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR