3-Fluorphenylmagnesium bromide 1 M in MeTHF (2-methyltetrahydrofuran)
Supplier: Thermo Scientific
H51168.AE
H51168.AP
H51168.AEEA
0
GBP
H51168.AE
H51168.AP
3-Fluorphenylmagnesium bromide 1 M in MeTHF (2-methyltetrahydrofuran)
3-Fluorphenylmagnesium bromide
CAS No.: 17318-03-5
Molecular Formula: C6H4BrFMg
Formula Weight: 199.31
Flash Point: -11°(12°F)
UN No.: 2924
MDL No.: MFCD00672007
Formula:
C₆H₄BrFMg Flash Pt: –11 °C (–12 °F) |
MDL Number:
MFCD00672007 CAS Number: 17318-03-5 UN: 2924 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...