N-(6-Methyl-2-pyridyl)thiourea ≥97%
Supplier: Thermo Scientific
|
Danger
|
H33884.03
H33884.06
H33884.03EA
55.4
GBP
H33884.03
H33884.06
N-(6-Methyl-2-pyridyl)thiourea ≥97%
N-(6-Methyl-2-pyridyl)thiourea
Purity: 97%
CAS No.: 49600-34-2
Molecular Formula: C7H9N3S
Formula Weight: 167.23
Melting Point: 193-197°
MDL No.: MFCD00462739
Formula:
C₇H₉N₃S MW: 167,23 g/mol Melting Pt: 193…197 °C Storage Temperature: Ambient |
MDL Number:
MFCD00462739 CAS Number: 49600-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...