Hexanophenone 98

Supplier: Thermo Fisher Scientific

A14712.06 A14712.14 A14712.22
A14712.06EA 24.3 GBP
A14712.06 A14712.14 A14712.22
Hexanophenone 98
Hexanophenone

Purity: 98% CAS No.: 942-92-7 Molecular Formula: C12H16O Formula Weight: 176.26 Flash Point: 110°(230°F) Refractive Index: 1.5105 Melting Point: 25-27° Boiling Point: 265° Beilstein Reference: 1908667 MDL No.: MFCD00009512

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Boiling Pt: 265 °C
Melting Pt: 25…27 °C
Density: 0.950
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009512
CAS Number: 942-92-7
EINECS: 213-394-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR