Vincristine sulphate, Millipore®
Supplier: Merck Millipore Calbiochem
677181-10EA
987
GBP
677181-10
Vincristine sulphate, Millipore®
Vincristine sulphate
Formula:
C₄₆H₅₈N₄O₁₄S MW: 923,04 g/mol |
MDL Number:
MFCD00084729 CAS Number: 2068-78-2 EINECS: 218-190-0 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...