2-Fluoro-m-toluic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Fluoro-3-methylbenzoic acid

A12280.03 A12280.06
A12280.03EA 19.4 GBP
A12280.03 A12280.06
2-Fluoro-m-toluic acid ≥98%
2-Fluoro-m-toluic acid

Purity: 98% CAS No.: 315-31-1 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Melting Point: 114-116° MDL No.: MFCD00042504

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00042504
CAS Number: 315-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR