2-Chloronicotinoyl chloride ≥98%
Supplier: Thermo Scientific
|
Danger
|
A11048.06
A11048.14
A11048.22
A11048.06EA
74.4
GBP
A11048.06
A11048.14
A11048.22
2-Chloronicotinoyl chloride ≥98%
2-Chloronicotinoyl chloride
Purity: 98+%
CAS No.: 49609-84-9
Molecular Formula: C6H3Cl2NO
Formula Weight: 176.00
Melting Point: 38-42°
Boiling Point: 98-100°/2mm
UN No.: 3261
MDL No.: MFCD00051677
Formula:
C₆H₃Cl₂NO MW: 176,00 g/mol Boiling Pt: 98…100 °C (2 mmHg) Melting Pt: 38…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051677 CAS Number: 49609-84-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...