4-(Benzyloxy)-2-hydroxybenzaldehyde ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
L15515.03
L15515.06
L15515.03EA
29
GBP
L15515.03
L15515.06
4-(Benzyloxy)-2-hydroxybenzaldehyde ≥99%
4-(Benzyloxy)-2-hydroxybenzaldehyde
Purity: 99%
CAS No.: 52085-14-0
Molecular Formula: C14H12O3
Formula Weight: 228.25
Melting Point: 77-80°
Beilstein Reference: 982995
Formula:
C₁₄H₁₂O₃ MW: 228.25 g/mol Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075698 CAS Number: 52085-14-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...