1-Naphthylacetamide ≥98%

Supplier: Thermo Fisher Scientific
Warning

B23986.14 B23986.22 B23986.36
B23986.14EA 27.3 GBP
B23986.14 B23986.22 B23986.36
1-Naphthylacetamide ≥98%
1-Naphthylacetamide

Purity: 98% CAS No.: 86-86-2 Molecular Formula: C12H11NO Formula Weight: 185.23 Melting Point: 176-178° MDL No.: MFCD00004047

Formula: C₁₂H₁₁NO
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00004047
CAS Number: 86-86-2
EINECS: 201-704-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR