2-Amino-5-pyrimidineboronic acid pinacol ester 96%

Supplier: Thermo Fisher Scientific
Warning

H54451.MD H54451.03
H54451.MDEA 0 GBP
H54451.MD H54451.03
2-Amino-5-pyrimidineboronic acid pinacol ester 96%
2-Amino-5-pyrimidineboronic acid pinacol ester

Purity: 96% CAS No.: 402960-38-7 Molecular Formula: C10H16BN3O2 Formula Weight: 221.06 Storage Temperature: 2°C to 8°C Melting Point: 206-208° MDL No.: MFCD06795685

Formula: C₁₀H₁₆BN₃O₂
Melting Pt: 206…208 °C
MDL Number: MFCD06795685
CAS Number: 402960-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR