5-Iodo-1-methyl-1H-pyrazole ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 5-Iodo-1-methylpyrazole

H37465.03 H37465.06
H37465.03EA 65.7 GBP
H37465.03 H37465.06
5-Iodo-1-methyl-1H-pyrazole ≥97%
5-Iodo-1-methyl-1H-pyrazole
Formula: C₄H₅IN₂
MW: 208,00 g/mol
Melting Pt: 78…81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11109327
CAS Number: 34091-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR