N-Phthaloylglycine ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: (1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetic acid

A15920.0B A15920.18 A15920.30
A15920.0BEA 415 GBP
A15920.0B A15920.18 A15920.30
N-Phthaloylglycine ≥98%
N-Phthaloylglycine

Purity: 98+% CAS No.: 4702-13-0 Molecular Formula: C10H7NO4 Formula Weight: 205.17 Melting Point: 194-198° Beilstein Reference: 184174

Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
Melting Pt: 194…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00005900
CAS Number: 4702-13-0
EINECS: 225-177-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR