3-Aminopropionitrile fumarate ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
B25440.06
B25440.14
B25440.06EA
30.7
GBP
B25440.06
B25440.14
3-Aminopropionitrile fumarate ≥98%
3-Aminopropionitrile fumarate
Purity: 98+%
CAS No.: 2079-89-2
Molecular Formula: (NH2CH2CH2CN)2.HO2CCH=CHCO2H
Formula Weight: 256.26
Odour: Odourless
Melting Point: ca 177° dec.
Beilstein Reference: 3573532
MDL No.: MFCD00078321
Formula:
C₁₀H₁₆N₄O₄ MW: 256.26 g/mol Melting Pt: ∼ 177 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00078321 CAS Number: 2079-89-2 EINECS: 218-208-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...