N-(5-Fluoro-2-pyridyl)thiourea ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
H33831.MD
H33831.03
H33831.MDEA
0
GBP
H33831.MD
H33831.03
N-(5-Fluoro-2-pyridyl)thiourea ≥97%
N-(5-Fluoro-2-pyridyl)thiourea
Purity: 97%
Molecular Formula: C6H6FN3S
Formula Weight: 171.19
Melting Point: 212-216°
MDL No.: MFCD17780187
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...