N-(5-Bromo-2-pyridyl)thiourea ≥97%

Supplier: Thermo Scientific
Danger

H34088.03 H34088.06
H34088.03EA 65.5 GBP
H34088.03 H34088.06
N-(5-Bromo-2-pyridyl)thiourea ≥97%
N-(5-Bromo-2-pyridyl)thiourea

Purity: 97% CAS No.: 31430-38-3 Molecular Formula: C6H6BrN3S Formula Weight: 232.10 Melting Point: 236-240° MDL No.: MFCD15525454

Formula: C₆H₆BrN₃S
MW: 232,10 g/mol
Melting Pt: 236…240 °C
Storage Temperature: Ambient
MDL Number: MFCD15525454
CAS Number: 31430-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR