N-(3,5-Dibromo-6-methyl-2-pyridyl)thiourea ≥97%
Supplier: Thermo Scientific
|
Danger
|
H34014.MD
H34014.03
H34014.MDEA
55.5
GBP
H34014.MD
H34014.03
N-(3,5-Dibromo-6-methyl-2-pyridyl)thiourea ≥97%
N-(3,5-Dibromo-6-methyl-2-pyridyl)thiourea
Purity: 97%
Molecular Formula: C7H7Br2N3S
Formula Weight: 325.02
Melting Point: 204-208°
MDL No.: MFCD22125032
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...