4-Nitrothiophenol 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-Nitrobenzene-1-thiol, 4-Nitrobenzenethiol, 1-Mercapto-4-nitrobenzene, p-Nitrothiophenol
H59240.06
H59240.14
H59240.06EA
91.8
GBP
H59240.06
H59240.14
4-Nitrothiophenol 96%
4-Nitrothiophenol
Formula:
C₆H₅NO₂S MW: 155.18 g/mol Melting Pt: 72…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007343 CAS Number: 1849-36-1 EINECS: 217-436-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...