α,3-Dibromo-2-fluorotoluene ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1-Bromo-3-(bromomethyl)-2-fluorobenzene, 3-Bromo-2-fluorobenzyl bromide
H32228.03
H32228.06
H32228.03EA
0
GBP
H32228.03
H32228.06
α,3-Dibromo-2-fluorotoluene ≥97%
α,3-Dibromo-2-fluorotoluene
Purity: 97%
CAS No.: 149947-16-0
Molecular Formula: C7H5Br2F
Formula Weight: 267.92
Boiling Point: 60°/0.1mm
UN No.: 3265
MDL No.: MFCD09842442
Formula:
C₇H₅Br₂F MW: 267.92 g/mol Boiling Pt: 60 °C (0.1 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD09842442 CAS Number: 149947-16-0 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...