2,4-Difluoro-3-methylbenzylamine 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
H32060.MD
H32060.03
H32060.MDEA
0
GBP
H32060.MD
H32060.03
2,4-Difluoro-3-methylbenzylamine 95%
2,4-Difluoro-3-methylbenzylamine
Purity: 98%
CAS No.: 847502-90-3
Molecular Formula: C8H9F2N
Formula Weight: 157.16
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Sensitivity: Air Sensitive
Appearance: Colourless to yellow
Odour: Amine-like
Boiling Point: 42/1mm
UN No.: 2735
MDL No.: MFCD08275293
Formula:
C₈H₉F₂N MW: 157.16 g/mol Boiling Pt: 42 °C (1 mmHg) |
MDL Number:
MFCD08275293 CAS Number: 847502-90-3 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...