4-Carboxy-2-fluorophenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Carboxy-2-fluorobenzeneboronic acid

H55857.06 H55857.14
H55857.06EA 569 GBP
H55857.06 H55857.14
4-Carboxy-2-fluorophenylboronic acid ≥97%
4-Carboxy-2-fluorophenylboronic acid

Purity: 97% CAS No.: 851135-07-4 Molecular Formula: C7H6BFO4 Formula Weight: 183.93 MDL No.: MFCD08458473

Formula: C₇H₆BFO₄
MW: 183,93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458473
CAS Number: 851135-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR