2-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥97%

Supplier: Thermo Scientific

Synonyms: 2-Ethoxycarbonylphenylboronic acid pinacol ester, 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

H30941.03 H30941.06
H30941.03EA 76.1 GBP
H30941.03 H30941.06
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥97%
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester

Purity: 97% CAS No.: 269409-99-6 Molecular Formula: C15H21BO4 Formula Weight: 276.14 Melting Point: 57-62° MDL No.: MFCD03453660

Formula: C₁₅H₂₁BO₄
MW: 276,136 g/mol
Melting Pt: 57…62 °C
Storage Temperature: Ambient
MDL Number: MFCD03453660
CAS Number: 269409-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR