4-Indoleboronic acid pinacol ester ≥97%

Supplier: Thermo Scientific

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

H52325.MD H52325.03
H52325.MDEA 48 GBP
H52325.MD H52325.03
4-Indoleboronic acid pinacol ester ≥97%
4-Indoleboronic acid pinacol ester

Purity: 97+% CAS No.: 388116-27-6 Molecular Formula: C14H18BNO2 Formula Weight: 243.11 MDL No.: MFCD08689896

Formula: C₁₄H₁₈BNO₂
MW: 243,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689896
CAS Number: 388116-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR