Vacuum ovens
Supplier: Thermo Fisher Scientific
BARN3618-6CEEA 9320 GBP
BARN3618-6CE BARN3618-1CE BARN3606-1CE BARN3608-6CE
Vacuum ovens
Ovens Vacuum Ovens
Achieve maximum temperatures of 220 °C with these Thermo Scientific™ vacuum ovens, while benefiting from two control configurations and display options. These ovens feature independent vacuum and purge needle valves. Radiant warm-wall heating system optimises uniformity and conserves chamber space for drying, curing, vacuum embedding and plating applications.
  • Hydraulic thermostat temperature control with dial thermometer (5 °C increments); medium and large units available with LED display (1 °C increments)
  • Polycarbonate safety shield protects door glass
  • Silicone door gasket and positive latch door maintain seal at all vacuum levels – use of vacuum grease is recommended
  • Vacuum and air lines are corrosion-resistant stainless-steel tubing for optimum chamber cleanliness and long-term performance
  • Two removable stacking aluminum shelves
  • Powder-coated heavy-gauge steel exterior for durability
  • Radiant warm-wall heating system helps optimize uniformity and conserve chamber space; 7,6 cm glass wool insulation prevents heat loss
Small 3606 models (not available in all countries): Top-mounted independent evacuation and venting vacuum fittings.
Medium and large 3608 and 3618 models: Front-mounted three-way valve for evacuation, venting and purging of inert gases (e.g. nitrogen or argon) with vacuum fittings on the front of the unit.