9-Methylacridine 96%
Supplier: Thermo Scientific
|
Warning
|
H33718.MD
H33718.03
H33718.MDEA
52.6
GBP
H33718.MD
H33718.03
9-Methylacridine 96%
9-Methylacridine
Formula:
C₁₄H₁₁N Melting Pt: 117…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143523 CAS Number: 611-64-3 EINECS: 210-272-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...