2-(2,3-Dichlorophenyl)thioacetamide ≥97%

Supplier: Thermo Scientific
Danger

H34058.03 H34058.MD
H34058.03EA 71.6 GBP
H34058.03 H34058.MD
2-(2,3-Dichlorophenyl)thioacetamide ≥97%
2-(2,3-Dichlorophenyl)thioacetamide

Purity: 97% CAS No.: 65183-01-9 Molecular Formula: C8H7Cl2NS Formula Weight: 220.12 Melting Point: 162-166° MDL No.: MFCD09757550

Formula: C₈H₇Cl₂NS
MW: 220,12 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD09757550
CAS Number: 65183-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR