3-(2,4-Dimethoxyphenyl)propionic acid 96%

Supplier: Thermo Scientific
Warning

Synonyms: 3-(2,4-Dimethoxyphenyl)propanoic acid

H34405.03 H34405.09
H34405.03EA 33.1 GBP
H34405.03 H34405.09
3-(2,4-Dimethoxyphenyl)propionic acid 96%
3-(2,4-Dimethoxyphenyl)propionic acid

Purity: 96% CAS No.: 22174-29-4 Molecular Formula: C11H14O4 Formula Weight: 210.23 Melting Point: 56-60° MDL No.: MFCD00598205

Formula: C₁₁H₁₄O₄
MW: 210,23 g/mol
Melting Pt: 56…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00598205
CAS Number: 22174-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR