Dimagnesium silicide ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific
|
Danger
|
12837.09
12837.18
12837.09EA
46.3
GBP
12837.09
12837.18
Dimagnesium silicide ≥99.5% (metals basis)
Dimagnesium silicide
Purity: 99.5%
CAS No.: 22831-39-6
Molecular Formula: Mg2Si
Formula Weight: 76.71
Physical Form: -20 Mesh Powder
Density: 2.0
Appearance: Blue
Melting Point: 1102°
Merck Reference: 14,5688
UN No.: 2624
MDL No.: MFCD00016202
Formula:
Mg₂Si Melting Pt: 1102 °C Density: 2,0 Storage Temperature: Ambient |
MDL Number:
MFCD00016202 CAS Number: 22831-39-6 EINECS: 245-254-5 UN: 2624 ADR: 4.3,II Merck Index: 14,5688 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...