4-Chloro-N-(4-fluorobenzyl)-2-methylaniline ≥97%

Supplier: Thermo Fisher Scientific
Danger

H56258.MD H56258.03
H56258.MDEA 0 GBP
H56258.MD H56258.03
4-Chloro-N-(4-fluorobenzyl)-2-methylaniline ≥97%
4-Chloro-N-(4-fluorobenzyl)-2-methylaniline
Formula: C₁₄H₁₃ClFN
MW: 249.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11119360
CAS Number: 1036628-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR