Potassium (4-acetylphenyl)trifluoroborate ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Potassium 4-acetylphenyltrifluoroborate, Potassium (4-acetylphenyl)trifluoroboranuide

H55845.03 H55845.06
H55845.03EA 0 GBP
H55845.03 H55845.06
Potassium (4-acetylphenyl)trifluoroborate ≥97%
Potassium 4-acetylphenyltrifluoroborate
Formula: C₈H₇BF₃KO
MW: 226.05 g/mol
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD07784376
CAS Number: 252726-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR