2-(4-Methylphenyl)-4-thiazolecarboxylic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-(p-Tolyl)thiazole-4-carboxylic acid

H54054.MD H54054.03
H54054.MDEA 24.1 GBP
H54054.MD H54054.03
2-(4-Methylphenyl)-4-thiazolecarboxylic acid ≥97%
2-(4-Methylphenyl)-4-thiazolecarboxylic acid

Purity: 97% CAS No.: 17228-99-8 Molecular Formula: C11H9NO2S Formula Weight: 219.26 Melting Point: 168-172° MDL No.: MFCD01936002

Formula: C₁₁H₉NO₂S
MW: 219,26 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD01936002
CAS Number: 17228-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR