Dicyclohexylchlorophosphine ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98+%
CAS No.: 16523-54-9
Molecular Formula: C12H22ClP
Formula Weight: 232.73
Flash Point:
Formula:
C₁₂H₂₂ClP Boiling Pt: 97 °C (0,1 mmHg) Density: 1,054 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014294 CAS Number: 16523-54-9 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...