4-Anilino-1-benzylpiperidine ≥98%

Supplier: Thermo Scientific

H27158.03 H27158.06
H27158.03EA 0 GBP
H27158.03 H27158.06
4-Anilino-1-benzylpiperidine ≥98%
4-Anilino-1-benzylpiperidine

Purity: 98% CAS No.: 1155-56-2 Molecular Formula: C18H22N2 Formula Weight: 266.38 MDL No.: MFCD00023738

Formula: C₁₈H₂₂N₂
MDL Number: MFCD00023738
CAS Number: 1155-56-2
EINECS: 214-583-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR